Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126658.1 | NP_001120130.1 | 11 - 12 | 1636 | 115 | |
GenBank mRNA | BC159054.1 | - | 11 - 12 | 1636 | 115 | |
CT030321.1 | - | 11 - 12 | 1736 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |