Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126658.1 | NP_001120130.1 | 3 - 4 | 401 | 114 | |
GenBank mRNA | BC159054.1 | - | 3 - 4 | 401 | 114 | |
CT030321.1 | - | 2 - 3 | 396 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |