Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nxt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126661.1 | NP_001120133.1 | 3 - 4 | 295 | 93 | |
GenBank mRNA | BC159063.1 | - | 3 - 4 | 295 | 93 | |
CT025236.2 | - | 3 - 4 | 248 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |