Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126677.1 | NP_001120149.1 | 1 - 2 | 510 | 70 | |
XM_012956449.2 | XP_012811903.1 | 1 - 2 | 569 | 70 | ||
GenBank mRNA | BC159142.1 | - | 1 - 2 | 510 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |