Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126677.1 | NP_001120149.1 | 3 - 4 | 712 | 70 | |
XM_012956449.2 | XP_012811903.1 | 3 - 4 | 771 | 70 | ||
GenBank mRNA | BC159142.1 | - | 3 - 4 | 712 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |