Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prelid3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126741.1 | NP_001120213.1 | 3 - 4 | 587 | 92 | |
XM_012964291.1 | XP_012819745.1 | 3 - 4 | 369 | 92 | ||
XM_012964292.1 | XP_012819746.1 | 3 - 4 | 274 | 92 | ||
XM_012964293.2 | XP_012819747.1 | 3 - 4 | 270 | 92 | ||
GenBank mRNA | BC159369.1 | - | 3 - 4 | 587 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |