Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prelid3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126741.1 | NP_001120213.1 | 6 - 7 | 854 | 70 | |
XM_012964291.1 | XP_012819745.1 | 6 - 7 | 636 | 70 | ||
XM_012964292.1 | XP_012819746.1 | 6 - 7 | 541 | 70 | ||
XM_012964293.2 | XP_012819747.1 | 6 - 7 | 537 | 70 | ||
XM_018094544.1 | XP_017950033.1 | 5 - 6 | 789 | 70 | ||
GenBank mRNA | BC159369.1 | - | 6 - 7 | 854 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |