Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126814.1 | NP_001120286.1 | 1 - 2 | 81 | 73 | |
GenBank mRNA | BC160555.1 | - | 1 - 2 | 81 | 73 | |
CR761688.2 | - | 1 - 2 | 162 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |