Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126957.1 | NP_001120429.1 | 13 - 14 | 1568 | 75 | |
XM_012966087.2 | XP_012821541.1 | 13 - 14 | 1646 | 75 | ||
XM_012966088.2 | XP_012821542.1 | 13 - 14 | 1647 | 75 | ||
XM_012966089.2 | XP_012821543.1 | 13 - 14 | 1643 | 75 | ||
GenBank mRNA | BC161184.1 | - | 13 - 14 | 1568 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |