Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsd3b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127029.1 | NP_001120501.1 | 5 - 6 | 557 | 83 | |
XM_012954311.2 | XP_012809765.1 | 4 - 5 | 549 | 83 | ||
XM_018097043.1 | XP_017952532.1 | 5 - 6 | 605 | 83 | ||
GenBank mRNA | BC161389.1 | - | 5 - 6 | 557 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |