Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127127.1 | NP_001120599.1 | 5 - 6 | 643 | 86 | |
GenBank mRNA | AB474592.1 | - | 4 - 5 | 549 | 86 | |
BC161634.1 | - | 5 - 6 | 643 | 86 | ||
CT030302.1 | - | 5 - 6 | 596 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |