Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127134.1 | NP_001120606.1 | 2 - 3 | 505 | 94 | |
GenBank mRNA | BC161735.1 | - | 2 - 3 | 505 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |