Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pigb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127136.1 | NP_001120608.1 | 7 - 8 | 928 | 107 | |
XM_018091939.1 | XP_017947428.1 | 7 - 8 | 826 | 107 | ||
XM_018091940.1 | XP_017947429.1 | 7 - 8 | 826 | 107 | ||
GenBank mRNA | BC161737.1 | - | 7 - 8 | 928 | 107 | |
CT025216.2 | - | 7 - 8 | 1104 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |