Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhfpl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127145.1 | NP_001120617.1 | 2 - 3 | 745 | 55 | |
XM_012956507.2 | XP_012811961.1 | 2 - 3 | 950 | 55 | ||
GenBank mRNA | BC161757.1 | - | 2 - 3 | 745 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |