Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dner | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122791.1 | NP_001116263.1 | 1129 | 80 | ||
GenBank mRNA | BC121488.1 | - | 1142 | 80 | ||
BC135484.1 | - | 1129 | 80 | |||
BC161016.1 | - | 1129 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |