Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126515.1 | NP_001119987.1 | 8 - 9 | 1029 | 75 | |
XM_012966046.2 | XP_012821500.1 | 9 - 10 | 1161 | 75 | ||
GenBank mRNA | BC158244.1 | - | 8 - 9 | 1029 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |