Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126517.1 | NP_001119989.1 | 7 - 8 | 1310 | 82 | |
XM_012956422.2 | XP_012811876.1 | 7 - 8 | 1121 | 82 | ||
XM_012956423.2 | XP_012811877.1 | 7 - 8 | 1128 | 82 | ||
XM_012956424.2 | XP_012811878.1 | 7 - 8 | 1101 | 82 | ||
XM_012956425.2 | XP_012811879.1 | 7 - 8 | 1057 | 82 | ||
XM_012956426.1 | XP_012811880.1 | 6 - 7 | 1038 | 82 | ||
XM_018091076.1 | XP_017946565.1 | 7 - 8 | 1056 | 82 | ||
XM_018091077.1 | XP_017946566.1 | 7 - 8 | 1055 | 82 | ||
XM_018091078.1 | XP_017946567.1 | 7 - 8 | 1348 | 82 | ||
GenBank mRNA | BC158248.1 | - | 7 - 8 | 1310 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |