Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cwc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126572.2 | NP_001120044.1 | 5 - 6 | 688 | 77 | |
GenBank mRNA | BC158381.1 | - | 5 - 6 | 690 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |