Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126653.1 | NP_001120125.1 | 306 | 65 | ||
GenBank mRNA | BC159034.1 | - | 306 | 65 | ||
CT027949.1 | - | 247 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |