Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gopc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126685.1 | NP_001120157.1 | 5 - 6 | 908 | 78 | |
XM_012962715.2 | XP_012818169.1 | 6 - 7 | 996 | 78 | ||
XM_012962716.2 | XP_012818170.1 | 6 - 7 | 993 | 78 | ||
XM_012962717.2 | XP_012818171.1 | 5 - 6 | 970 | 78 | ||
XM_012962718.2 | XP_012818172.1 | 5 - 6 | 967 | 78 | ||
GenBank mRNA | BC159156.1 | - | 5 - 6 | 908 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |