Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cuedc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126757.1 | NP_001120229.1 | 6 - 7 | 547 | 104 | |
XM_018095262.1 | XP_017950751.1 | 6 - 7 | 581 | 104 | ||
GenBank mRNA | BC160403.1 | - | 6 - 7 | 547 | 104 | |
CR848581.1 | - | 6 - 7 | 534 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |