Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cln6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126758.1 | NP_001120230.1 | 6 - 7 | 742 | 113 | |
GenBank mRNA | BC160405.1 | - | 6 - 7 | 742 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |