Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126807.1 | NP_001120279.1 | 4 - 5 | 1144 | 70 | |
GenBank mRNA | BC160543.1 | - | 4 - 5 | 1144 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |