Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dipk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126932.1 | NP_001120404.1 | 2 - 3 | 1196 | 96 | |
GenBank mRNA | BC161119.1 | - | 2 - 3 | 1196 | 96 | |
CT025359.2 | - | 2 - 3 | 1164 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |