Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126935.1 | NP_001120407.1 | 8 - 9 | 1109 | 84 | |
XM_012962732.2 | XP_012818186.1 | 8 - 9 | 1189 | 84 | ||
XM_012962733.2 | XP_012818187.1 | 8 - 9 | 1255 | 84 | ||
XM_012962734.2 | XP_012818188.1 | 8 - 9 | 1153 | 84 | ||
XM_012962735.2 | XP_012818189.1 | 9 - 10 | 1660 | 84 | ||
XM_012962736.2 | XP_012818190.1 | 9 - 10 | 1657 | 84 | ||
XM_012962737.1 | XP_012818191.1 | 8 - 9 | 1146 | 84 | ||
XM_012962738.2 | XP_012818192.2 | 6 - 7 | 722 | 84 | ||
GenBank mRNA | BC161123.1 | - | 8 - 9 | 1109 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |