Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hs2st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126991.1 | NP_001120463.1 | 3 - 4 | 620 | 108 | |
XM_018092933.1 | XP_017948422.1 | 3 - 4 | 708 | 108 | ||
GenBank mRNA | BC161283.1 | - | 3 - 4 | 620 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |