Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdelc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127103.1 | NP_001120575.1 | 5 - 6 | 1144 | 70 | |
GenBank mRNA | BC161552.1 | - | 5 - 6 | 1144 | 70 | |
BC167365.1 | - | 5 - 6 | 1144 | 70 | ||
BC168093.1 | - | 5 - 6 | 1144 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |