Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tcp11l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127106.1 | NP_001120578.1 | 5 - 6 | 834 | 116 | |
GenBank mRNA | BC161560.1 | - | 5 - 6 | 834 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |