Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127148.1 | NP_001120620.1 | 11 - 12 | 1056 | 104 | |
GenBank mRNA | BC161764.1 | - | 11 - 12 | 1056 | 104 | |
CU075325.1 | - | 11 - 12 | 1060 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |