Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127156.1 | NP_001120628.1 | 9 - 10 | 1612 | 74 | |
GenBank mRNA | BC161780.1 | - | 9 - 10 | 1612 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |