Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stx10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127158.1 | NP_001120630.1 | 4 - 5 | 659 | 90 | |
GenBank mRNA | BC161783.1 | - | 4 - 5 | 659 | 90 | |
CT025172.2 | - | 4 - 5 | 386 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |