Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkmyt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127159.1 | NP_001120631.1 | 4 - 5 | 1058 | 86 | |
XM_012970105.2 | XP_012825559.1 | 4 - 5 | 1143 | 86 | ||
XM_012970106.2 | XP_012825560.1 | 4 - 5 | 1109 | 86 | ||
XM_012970107.2 | XP_012825561.1 | 4 - 5 | 1920 | 86 | ||
GenBank mRNA | BC161784.1 | - | 4 - 5 | 1058 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |