Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100145817 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018091006.1 | XP_017946495.1 | 3 - 4 | 315 | 84 | |
GenBank mRNA | EF513165.1 | - | 1 - 2 | 128 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Xenopus tropicalis Probe spans exons |