Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127168.1 | NP_001120640.1 | 13 - 14 | 2545 | 105 | |
XM_012958613.2 | XP_012814067.1 | 14 - 15 | 2710 | 105 | ||
XM_012958614.2 | XP_012814068.1 | 14 - 15 | 2636 | 105 | ||
GenBank mRNA | BC166189.1 | - | 13 - 14 | 2545 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |