Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nomo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127971.1 | NP_001121443.1 | 13 - 14 | 1603 | 82 | |
GenBank mRNA | BC166195.1 | - | 13 - 14 | 1603 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |