Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nomo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127971.1 | NP_001121443.1 | 24 - 25 | 2939 | 88 | |
GenBank mRNA | BC166195.1 | - | 24 - 25 | 2939 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |