Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127973.2 | NP_001121445.2 | 10 - 11 | 1311 | 95 | |
GenBank mRNA | BC166198.1 | - | 1300 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |