Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nuak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128027.1 | NP_001121499.1 | 6 - 7 | 790 | 69 | |
GenBank mRNA | BC166319.1 | - | 6 - 7 | 790 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |