Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128030.1 | NP_001121502.1 | 5 - 6 | 603 | 101 | |
GenBank mRNA | BC166331.1 | - | 5 - 6 | 603 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |