Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nop9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128031.1 | NP_001121503.1 | 2 - 3 | 278 | 62 | |
GenBank mRNA | BC166335.1 | - | 2 - 3 | 278 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |