Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128046.1 | NP_001121518.1 | 5 - 6 | 556 | 99 | |
XM_018093802.1 | XP_017949291.1 | 5 - 6 | 589 | 99 | ||
GenBank mRNA | BC166373.1 | - | 5 - 6 | 556 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |