Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128640.1 | NP_001122112.1 | 7 - 8 | 3404 | 86 | |
XM_012964011.2 | XP_012819465.1 | 6 - 7 | 3629 | 86 | ||
XM_012964013.2 | XP_012819467.1 | 6 - 7 | 3670 | 86 | ||
XM_012964015.2 | XP_012819469.1 | 5 - 6 | 3408 | 86 | ||
XM_012964018.2 | XP_012819472.1 | 5 - 6 | 3534 | 86 | ||
GenBank mRNA | BC167678.1 | - | 7 - 8 | 3404 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |