Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130221.1 | NP_001123693.1 | 4 - 5 | 649 | 90 | |
XM_012956966.2 | XP_012812420.1 | 4 - 5 | 876 | 90 | ||
GenBank mRNA | BC167275.1 | - | 4 - 5 | 649 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |