Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mcu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130227.1 | NP_001123699.1 | 2 - 3 | 379 | 75 | |
GenBank mRNA | BC167298.1 | - | 2 - 3 | 379 | 75 | |
CR942334.2 | - | 2 - 3 | 373 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |