Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130369.1 | NP_001123841.1 | 5 - 6 | 825 | 91 | |
XM_012970169.2 | XP_012825623.1 | 5 - 6 | 1387 | 91 | ||
GenBank mRNA | BC167689.1 | - | 5 - 6 | 825 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |