Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exosc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130379.1 | NP_001123851.1 | 7 - 8 | 494 | 68 | |
GenBank mRNA | BC167725.1 | - | 7 - 8 | 494 | 68 | |
CR942534.2 | - | 7 - 8 | 485 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |