Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130383.1 | NP_001123855.1 | 3 - 4 | 216 | 70 | |
GenBank mRNA | BC167734.1 | - | 3 - 4 | 216 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |