Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134803.1 | NP_001128275.1 | 8 - 9 | 1410 | 70 | |
GenBank mRNA | BC087987.1 | - | 8 - 9 | 1413 | 70 | |
BC135297.1 | - | 8 - 9 | 1410 | 70 | ||
BC168783.1 | - | 8 - 9 | 1410 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |