Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134810.1 | NP_001128282.1 | 2 - 3 | 430 | 63 | |
XM_012953823.2 | XP_012809277.1 | 2 - 3 | 960 | 63 | ||
XM_012953824.2 | XP_012809278.1 | 2 - 3 | 1143 | 63 | ||
XM_018096085.1 | XP_017951574.1 | 2 - 3 | 600 | 63 | ||
GenBank mRNA | BC168574.1 | - | 2 - 3 | 430 | 63 | |
CR762100.2 | - | 2 - 3 | 406 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |