Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134810.1 | NP_001128282.1 | 3 - 4 | 576 | 118 | |
XM_012953823.2 | XP_012809277.1 | 3 - 4 | 1106 | 118 | ||
XM_012953824.2 | XP_012809278.1 | 3 - 4 | 1289 | 118 | ||
XM_012953825.2 | XP_012809279.1 | 4 - 5 | 1208 | 118 | ||
XM_018096085.1 | XP_017951574.1 | 3 - 4 | 746 | 118 | ||
GenBank mRNA | BC168574.1 | - | 3 - 4 | 576 | 118 | |
CR762100.2 | - | 3 - 4 | 552 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |